ContactUsLinkComponent
Demountable Quartz Torch, 3-Slot, for Optima 2x00/4x00/5x00/7x00 DV, PerkinElmer
Demountable Quartz Torch, 3-Slot, for Optima 2x00/4x00/5x00/7x00 DV, PerkinElmer
Catalog # 10136-320
Supplier:  PerkinElmer
undefined
Demountable Quartz Torch, 3-Slot, for Optima 2x00/4x00/5x00/7x00 DV, PerkinElmer
Catalog # 10136-320
Supplier:  PerkinElmer
Supplier Number:  N0772005
Specifications
  • Description:
    3-slot demountable quartz torch for Optima
  • Cat. No.:
    10136-320
  • Supplier no.:
    N0772005
Specifications
About this item
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flows. This style torch is recommended for use in organic analysis.
  • Demountable design permits quick changing of the torch body only, saving time and money
  • Recommended for use in organic analyses
This 3-Slot quartz torch is compatible with the Optima™ 2x00 DV, 4x00 DV, 5x00 DV, and 7x00 DV ICP-OES series PerkinElmer instruments.