Order Entry
Puerto Rico
ContactUsLinkComponent
Drum Plug Sockets for Round and Hex Head Drums, Labelmaster
Drum Plug Sockets for Round and Hex Head Drums, Labelmaster
Catalog # 103258-084
Supplier:  Label Master
Drum Plug Sockets for Round and Hex Head Drums, Labelmaster
Catalog # 103258-084
Supplier:  Label Master
Supplier Number:  KSOK234

Specifications

  • Material:
    Steel alloy
  • Cat. No.:
    103258-084
  • Weight:
    2 lbs.
  • Supplier no.:
    KSOK234

Specifications

About this item

Get the proper fit using Drum Plug Sockets for Round and Hex Head Drums.

  • Remove virtually all screw-in drum plugs quickly
  • Use these sockets with ½" drive ratchet or impact wrench
  • Drum accessory fits 2" and ¾" round or hex head steel plugs

Designed with a spring-loaded inner wrench that handles smaller plugs, these sockets recess automatically when removing large plugs.