Hazmat Shipping Paper Tab, Labelmaster
101377-576
- Description:Hazmat shipping paper tab
- Dimensions:19.1×50.8 mm (0.75×2")
- Material:Fluorescent red paper
- Thickness:7.1 mil.
- Environmentally Preferable:
- Cat. No.:101377-576
- Legend:Hazardous Materials
- Supplier no.:LT196U
Hazmat Shipping Paper tabs are high-visibility shipping tabs that instantly command attention wherever they were used.
- Black lettering on bright red background
- Can also be used to flag all hazardous materials shipping forms
- Alert shipping and receiving personnel to special handling instructions
When hazmat shipping forms must be visible and have to be noticed, use hazmat shipping paper tabs to increase DOT shipping form visibility.