Aluminum Placard 'Dangerous When Wet', Labelmaster
Catalog # 103508-348
Supplier: Label Master
Specifications
- Description:Aluminum placard
- Dimensions:273.1×273.1 mm (10.75×10.75")
- Material:Aluminum alloy
- Environmentally Preferable:
- Cat. No.:103508-348
- Legend:Dangerous When Wet
Specifications
About this item
Designed to meet DOT specifications for color and size, these Hazard Class 4 Dangerous When Wet Placards may be used domestically as well as internationally.
- Long lasting use
- Constructed of a corrosion-resistant aluminum alloy
- Will withstand the toughest road and weather conditions
Hazard Class 4 Dangerous When Wet Worded Placards are required by 49 CFR 172.500 for highway, rail, and water shipments of hazardous materials. The acrylic paint finish is baked on at 400° for superior durability. This can be used in Slide master holders.