Order Entry
Puerto Rico
ContactUsLinkComponent
International Infectious Substance Labels, Labelmaster
International Infectious Substance Labels, Labelmaster
Catalog # 101378-026
Supplier:  Label Master
International Infectious Substance Labels, Labelmaster
Catalog # 101378-026
Supplier:  Label Master
Supplier Number:  SLR17

Specifications

  • Description:
    Warning labels
  • Dimensions:
    100×100 mm (3.94×3.94")
  • Material:
    PVC-free film
  • Thickness:
    7.1 mil.
  • Cat. No.:
    101378-026
  • Legend:
    Infectious Substance
  • Supplier no.:
    SLR17

Specifications

About this item

Help take the guessing out of shipping label solutions with International Infectious Substance Labels, revised to meet the design requirements of 49 CFR 172.407.

  • Printed with light-fast inks for fade resistance
  • Adhesives formulated for use in various climates and environments
  • Strong holds on corrugated boxes
  • Once affixed, labels won't fade or fall off
  • Outstanding chemcial and abrasion resistance

In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.