Order Entry
Puerto Rico
ContactUsLinkComponent
Labels 'Hot', Labelmaster
Labels 'Hot', Labelmaster
Catalog # 101377-478
Supplier:  Label Master
Labels 'Hot', Labelmaster
Catalog # 101377-478
Supplier:  Label Master
Supplier Number:  B401

Specifications

  • Description:
    Preprinted labels
  • Dimensions:
    50.8×63.5 mm (2×2.5")
  • Material:
    Fluorescent red paper
  • Thickness:
    7.1 mil.
  • Cat. No.:
    101377-478
  • Packaging:
    On roll
  • Legend:
    Hot!
  • Supplier no.:
    B401

Specifications

About this item

Get a message across loud and clear to shipping personnel with Hot labels.

  • Ultra-bold graphics
  • Fluorescent red ink
  • Die cut on heavyweight paper
  • Strong, permanent adhesive

These high-visibility ColorBrite™ warehouse labels command instant attention wherever they’re used.