Order Entry
Puerto Rico
ContactUsLinkComponent
Labels 'USA Flag', Labelmaster
Labels 'USA Flag', Labelmaster
Catalog # 101377-152
Supplier:  Label Master
Labels 'USA Flag', Labelmaster
Catalog # 101377-152
Supplier:  Label Master
Supplier Number:  USA5

Specifications

  • Description:
    Preprinted labels
  • Dimensions:
    47.6×69.9 mm (1.88×2.75")
  • Material:
    Semi-gloss paper
  • Thickness:
    5.85 mil.
  • Cat. No.:
    101377-152
  • Packaging:
    On roll
  • Legend:
    Made In The U.S.A.
  • Supplier no.:
    USA5

Specifications

About this item

Put your American pride on display with Made in the USA Flag labels.

  • Printed on heavyweight coated paper
  • Red, white, and blue