PCB Labels, Labelmaster
Catalog # 101377-722
Supplier: Label Master
Specifications
- Description:PCB labels
- Dimensions:25.4×50.8 mm (1×2")
- Material:PVC-free film
- Thickness:7.1 mil.
- Cat. No.:101377-722
- Core size:3"
- Legend:No PCBs
- Supplier no.:PCNL
Specifications
About this item
These hazardous substance labels feature the printed warning: No PCBs.
- Pressure-sensitive adhesive backing
- Automotive-grade ink pigments
- Specially formulated adhesives
- Constructed of rugged, flexible material
The environmental labels are printed with fade-resistant inks on PVC-free film to provide all-weather durability.