PG III, Labels, Labelmaster
Catalog # 101376-334
Supplier: Label Master
Specifications
- Description:Labels
- Dimensions:100×100 mm (3.94×3.94")
- Material:Semi-gloss paper
- Thickness:5.85 mil.
- Cat. No.:101376-334
- Core size:3"
- Legend:PG III
- Supplier no.:HML32
Specifications
About this item
Help take the guessing out of shipping label solutions with Worded PG III labels.
- Revised to meet the design requirements of 49 CFR 172.407
- Complies with the directives of the Department of Transportation
- Meet the design and durability standards of 49 CFR, ICAO, and other international air regulations
These Hazard Class 6 labels are printed with light-fast inks for fade resistance, and adhesives are formulated for use in various climates and environments to ensure strong holds on corrugated boxes. Once affixed, they won't fade or fall off. They also feature outstanding chemical and abrasion resistance. White and black, these paper labels are sized at 100×100mm (4×4"), and sold in rolls of 500.