To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Style 258 peek
- Tip opening:1.5 mm
- Cat. No.:76376-056
- Overall length:125 mm
- Tip width:10 mm
- Tip length:65 mm
Specifications
About this item
Swiss made tweezers have tips with 30% carbon fibers. The tips are ESD safe, anti-magnetic, anti acid as well as temperature resistance up to 250 °C (short exposure up to 300 °C). Can be sterilized at 143 °C. Often used in the disc drive industry and for cer