Orders LinkContactUsLinkComponent
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Certificates
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
13 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Conventional forceps made of premium grade, hardened stainless steel with a protective coating for extra durability and protection from rust and corrosion.
  • Satin-matte finish reduces eye-fatiguing glare
  • Available in a variety of length, tip, and handle styles
Curved styles are well suited for retreiving specimens from the inside perimeter of transport jars and tissue separation in the water bath.
Product Family Options
Paddle Handle Forceps
Product Information
  • Overall length
    Tip length
    Tip configuration
    Availability
    Price
  • 76376-004
    Ergonomic forceps, paddle handle
    Ergonomic forceps, paddle handle
    178 mm
    3 mm
    Smooth tips
    Log in for availability
    Log in for availability
    Each
    Each - $209.85
    $209.85
    Each
       $209.85
    Supplier Number: 77937-44
  • 76376-006
    Ergonomic forceps, paddle handle
    Ergonomic forceps, paddle handle
    178 mm
    3 mm
    Serrated tips
    Log in for availability
    Log in for availability
    Each
    Each - $236.51
    $236.51
    Each
       $236.51
    Supplier Number: 77937-45
  • 76376-008
    Ergonomic forceps, paddle handle
    Ergonomic forceps, paddle handle
    178 mm
    3 mm
    Tips with 1×2 teeth
    Log in for availability
    Log in for availability
    Each
    Each - $227.86
    $227.86
    Each
       $227.86
    Supplier Number: 77937-46
  • 76376-010
    Ergonomic forceps, paddle handle
    Ergonomic forceps, paddle handle
    178 mm
    1.5 mm
    Smooth tips
    Log in for availability
    Log in for availability
    Each
    Each - $225.00
    $225.00
    Each
       $225.00
    Supplier Number: 77937-47
  • 76376-012
    Ergonomic forceps, paddle handle
    Ergonomic forceps, paddle handle
    178 mm
    1.5 mm
    Serrated tips
    Log in for availability
    Log in for availability
    Each
    Each - $225.60
    $225.60
    Each
       $225.60
    Supplier Number: 77937-48
  • 76376-014
    Ergonomic forceps, paddle handle
    Ergonomic forceps, paddle handle
    178 mm
    Needle tip
    Log in for availability
    Log in for availability
    Each
    Each - $214.59
    $214.59
    Each
       $214.59
    Supplier Number: 77937-49
  • 76376-016
    Ergonomic forceps, paddle handle
    Ergonomic forceps, paddle handle
    178 mm
    1.5 mm
    Curved serrated tips
    Log in for availability
    Log in for availability
    Each
    Each - $222.76
    $222.76
    Each
       $222.76
    Supplier Number: 77937-54
  • 76376-018
    Ergonomic forceps, paddle handle
    Ergonomic forceps, paddle handle
    178 mm
    1.5 mm
    Curved smooth tips
    Log in for availability
    Log in for availability
    Each
    Each - $355.31
    $355.31
    Each
       $355.31
    Supplier Number: 77937-55
Ergonomic Forceps
Product Information
  • Overall length
    Tip length
    Tip configuration
    Availability
    Price
  • 76376-020
    Ergonomic forceps
    Ergonomic forceps
    140 mm
    1.5 mm
    Straight smooth tips
    Log in for availability
    Log in for availability
    Each
    Each - $216.42
    $216.42
    Each
       $216.42
    Supplier Number: 77937-56
  • 76376-022
    Ergonomic forceps
    Ergonomic forceps
    140 mm
    1.5 mm
    Curved smooth tips
    Log in for availability
    Log in for availability
    Each
    Each - $316.76
    $316.76
    Each
       $316.76
    Supplier Number: 77937-57
  • 76376-024
    Ergonomic forceps
    Ergonomic forceps
    140 mm
    1.5 mm
    Curved serrated tips
    Log in for availability
    Log in for availability
    Each
    Each - $335.88
    $335.88
    Each
       $335.88
    Supplier Number: 77937-58
  • 76376-026
    Ergonomic forceps
    Ergonomic forceps
    140 mm
    1.5 mm
    Straight serrated tips
    Log in for availability
    Log in for availability
    Each
    Each - $327.32
    $327.32
    Each
       $327.32
    Supplier Number: 77937-59
  • 76376-028
    Ergonomic forceps
    Ergonomic forceps
    140 mm
    1.5 mm
    Needle serrated tips
    Log in for availability
    Log in for availability
    Each
    Each - $300.20
    $300.20
    Each
       $300.20
    Supplier Number: 77937-60