ContactUsLinkComponent
Snap Cap with Septa, Narrow Mouth, DWK Life Sciences
Snap Cap with Septa, Narrow Mouth, DWK Life Sciences
Catalog # 10216-868
undefined
Snap Cap with Septa, Narrow Mouth, DWK Life Sciences
Catalog # 10216-868
Supplier Number:  73826-2
Specifications
  • Color:
    Clear
  • Description:
    Autosampler snap cap
  • Cat. No.:
    10216-868
  • Supplier no.:
    37826-2
Specifications
About this item
Snap cap with no slit PTFE/silicone septa.
  • Autoclavable