ContactUsLinkComponent
GemCone Low-Flow Nebulizer for Optima and Avio ICP-OES, PerkinElmer
GemCone Low-Flow Nebulizer for Optima and Avio ICP-OES, PerkinElmer
Catalog # 10138-112
Supplier:  PerkinElmer
undefined
GemCone Low-Flow Nebulizer for Optima and Avio ICP-OES, PerkinElmer
Catalog # 10138-112
Supplier:  PerkinElmer
Supplier Number:  N0770358
Specifications
  • Description:
    GemCone Low-flow nebulizer for Optima and Avio
  • Environmentally Preferable:
  • Cat. No.:
    10138-112
  • Supplier no.:
    N0770358
Specifications
About this item
Low-flow GemCone nebulizers permit lower nebulizer gas flow rates, useful for spectral lines with high excitation energies providing a more robust plasma.
  • Compatible with PerkinElmer Optima® 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV, 8x00, 4300 V, 5300 V, 7300 V, and Avio® 200/500 ICP-OES series instruments
  • The nebulizer is optimized for low nebulizer gas flow for use with cyclonic spray chambers
  • Requires end cap (N0680504) for use with Ryton™ Scott spray chambers
GemCone Nebulizer is included in the Organics Sample Introduction System for Avio 200 (N0790611), Organics Sample Introduction System for Avio 500 (N0810609), Quick-Change Torch Module for Oil Analysis for Optima 4300V/5300V/7300V (N0770910), Organics Kit for Avio 200/500 (N0790610), Organics Kit for Optima 8×00 (N0780608), Organics Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770420), and the Organics Kit for Optima 4300 V/5300 V/7300 V (N0770732).