ContactUsLinkComponent
Spectroquant® Test Kits, Surfactants (Cell Test), MilliporeSigma
Spectroquant® Test Kits, Surfactants (Cell Test), MilliporeSigma
Catalog # EM1.02552.0001
Supplier:  MilliporeSigma
CAS Number:  
undefined
Spectroquant® Test Kits, Surfactants (Cell Test), MilliporeSigma
Catalog # EM1.02552.0001
Supplier:  MilliporeSigma
Supplier Number:  1.02552.0001
CAS Number:  
Specifications
  • Chemical method:
    Methylene blue
  • Description:
    Surfactants (anionic) cell test
  • Cat. No.:
    EM1.02552.0001
  • Range:
    0.05 - 2.00 mg/l SDSA
  • For unit:
    Prove, Nova 60, Multy
Specifications
About this item
Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.