Red Beam LaserShields, NoIR Laser Company
Supplier: NOIR MEDICAL
Certificates
About this item
CE and ANSI certified laser protection eyewear.
- Lightweight polycarbonate
Available in sporty frame #52, fit-over frame #53, or goggle frame #60.
Certifications: CE Certified LaserShields meet ANSI Z136.1, EN 207/EN 208/EN 60825, and Z87.1 for impact.
Ordering information: Hard-shell case, cleaning cloth, and strap included.
Frequently Bought Together