Order Entry
United States
ContactUsLinkComponent
KIMBLE® MICROFLEX® Erlenmeyer Flasks with Screw Cap, Graduated, DWK Life Sciences
KIMBLE® MICROFLEX® Erlenmeyer Flasks with Screw Cap, Graduated, DWK Life Sciences
Catalog # KT749400-0025
undefined
KIMBLE® MICROFLEX® Erlenmeyer Flasks with Screw Cap, Graduated, DWK Life Sciences
Catalog # KT749400-0025
Supplier Number:  749400-0025

Specifications

  • Cap type:
    Screw cap
  • Graduation Interval:
    5 ml
  • Graduation Range:
    10 - 25 ml
  • Capacity:
    25 ml
  • Environmentally Preferable:
  • Cat. No.:
    KT749400-0025
  • Thread:
    20 - 400
  • Supplier no.:
    749400-0025

Specifications

About this item

Graduated flask useful for sample collection, derivatization reactions, extractions, sample storage, micro mixing and more.

Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Delivery: Supplied with open top phenolic cap and PTFE-lined silicone septum.

Replacement Cap

Product Information