ContactUsLinkComponent
Claisen Adapter, Threaded, DWK Life Sciences
Claisen Adapter, Threaded, DWK Life Sciences
Catalog # KT747110-0000
undefined
Claisen Adapter, Threaded, DWK Life Sciences
Catalog # KT747110-0000
Supplier Number:  747110-0000
Specifications
  • Description:
    Claisen Adapter, 13-425
  • Height:
    50 mm
  • Cat. No.:
    KT747110-0000
  • Supplier no.:
    747110-0000
Specifications
About this item
This adapter converts single-neck flasks or vials to three-neck vessels for chemical reactions such as Grignard reactions.
  • Allows easy, vacuum-tight connections of syringes for liquid addition, thermometers or gas inlet tubes
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.