Reaction Flask Clamp, DWK Life Sciences
Catalog # 10135-804
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Reaction flask clamp
- Cat. No.:10135-804
- Flange I.D.:4"
- Supplier no.:613750-0000
Specifications
About this item
Aluminum construction to connect tops to flasks.
Wing nuts or thumb screws are spaced to assure uniform compression for sealing the system.