Distillation Adapters, 105° Bend, with Vacuum take-off and Outer Hose Connection, DWK Lifesciences
KT276405-0000
- Description:Extended 105 ° bent distillation adapters with vacuum take-off and outer hose connection
- Height:100 mm
- Cat. No.:KT276405-0000
- [ST] joint size:14/20
- Supplier no.:276405-0000
These extended 105° bent distillation adapters with vacuum take-off and outer hose connection.
- Barbed hose connection
- Joints are the same size
Standard Taper inner joint at the bottom and Standard Taper outer joint at the top.
: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.