Sample Vials, Amber, with Open-Top Screw Cap, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Amber screw thread sample vials are manufactured from 51 expansion borosilicate amber glass conforming to USP Type I requirements.
- Open-top white polypropylene closures
- PTFE-faced silicone septa attached
89201-844 features 0.005" PTFE-faced septa on 0.120" silicone liner.
89201-846 features 0.0055" PTFE-faced septa on 0.06" silicone liner.
Packaging: Vials packaged in corrugated trays with partitions.