ContactUsLinkComponent
SPERM-CHEX® Sperm Count Control, Streck
SPERM-CHEX® Sperm Count Control, Streck
Catalog # 10017-776
CAS Number:  
undefined
SPERM-CHEX® Sperm Count Control, Streck
Catalog # 10017-776
Supplier Number:  230202
CAS Number:  
Specifications
  • Description:
    SPERM-CHEX® Sperm count control
  • Size:
    2×300 µl (level 1, level 2)
  • Cat. No.:
    10017-776
  • Supplier no.:
    230202
Specifications
About this item
A procedural control with two levels of stabilized sperm cells to validate manual sperm count processes.
  • Comprehensive
  • Two clinically significant levels of control
  • The only sperm count control on the market that contains sperm cells
  • Mimics a patient sample
The control has a 12-month closed-vial stability and a 42-day open-vial stability.