A1c-Cellular® Linearity Control, Streck
10185-902
:
- Description:A1c-Cellular® Linearity
- Size:5×2 ml (levels 1-5)
- Cat. No.:10185-902
- Supplier no.:285536
A linearity product with instrument-specific target values and ranges for immediate results.
- Ready-to-use liquid control
- For immunoassay and ionic exchange HPLC methodologies
- Assayed on top 10 HbA1c analyzers
- Tests the entire HbA1c procedure, including the lysing of red blood cells
- Available in a 5-level set
Available in plastic cap-pierceable vials for analyzer autosampling. Only available HbA1c linearity material containing intact red blood cells.