To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Oxytetracycline-glucose-yeast extract agar
- Size:500 g
- Cat. No.:EM1.05978.0500
- Supplier no.:1.05978.0500
Oxytetracycline-glucose-yeast extract agar is used for the selective isolation and enumeration of yeasts and moulds in foods.
- pH: 6.6 ±0.2 at 25 °C
The base medium allows good growth of yeasts and moulds. Oxytetracycline inhibits the growth of bacteria.