Order Entry
Puerto Rico
ContactUsLinkComponent
Acetonitrile with 0,1% (v/v) acetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Flammable
Irritant
Acetonitrile with 0,1% (v/v) acetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Catalog # EM1.59004.2500
Supplier:  MilliporeSigma
   Safer Human and Environmental Health
CAS Number:  
Flammable
Irritant
Acetonitrile with 0,1% (v/v) acetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Catalog # EM1.59004.2500
Supplier:  MilliporeSigma
Supplier Number:  1.59004.2500
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2.5 L
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental Certification:
    Chem21 Solvent selection guide recommended by default
  • Cat. No.:
    EM1.59004.2500

Specifications

About this item

 :
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Storage Temperature:Ambient
UN:1648
ADR:3,
 :M-Clarity™ quality level = MQ100
Flash Pt:2 °C
 :GHS02
 :GHS07
 :64-19-7