Urinalysis Transport Tube with Tartaric (BTA) Tablet, Trinity Biotech
Supplier: Avantor
Certificates
About this item
The conical Urinalysis Transport Tubes offer the advantage of transporting urinalysis specimens without refrigeration.
- Dip stick and microscopic examination can be performed up to 72 hours at RT
- Tubes maintain acid pH and prevent bacterial proliferation
- Pathological components are not obscured
- 24 months shelf life
The preservative maintains the pH and prevents bacterial proliferation, thereby preventing false positive readings for occult blood and protein. Also, the identification of formed elements, such as cells and casts, which are enhanced while in vitro crystal formation is prevented.
The yellow and blue top urinalysis transport tubes contain the same BTA tablet, but the colored tops can be used for easy visual specimen separation between departments.
Packaging: Case size is 12×9×15 and weighs 8.5 lbs.
Caution: This product is for In vitro use only. Do not ingest the preservative tablet or powder. Do not use the specimen if the level of urine is less than mid-point of the urine preservative transport container. Autoclave all contaminated articles after use.
Frequently Bought Together