Order Entry
United States
ContactUsLinkComponent
Calibration Verification T-Uptake, Audit MicroControls, Inc.
Calibration Verification T-Uptake, Audit MicroControls, Inc.
Catalog # 97068-938
CAS Number:  
undefined
Calibration Verification T-Uptake, Audit MicroControls, Inc.
Catalog # 97068-938
Supplier Number:  K851M-5
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Cal Ver LQ T-Uptake for Tosoh A1A
  • Size:
    5×1 ml
  • Environmentally Preferable:
  • Cat. No.:
    97068-938
  • Stability:
    10 days when stored at 2...8 °C
  • Analyte:
    T-Uptake
  • Type:
    Liquid
  • Supplier no.:
    K851M-5

Specifications

About this item

T-Uptake calibration verification set is assayed quality control material consisting of five levels of human serum proteins.

T-Uptake calibration verification set is a quality control material intended for use in the quantitative verification of calibration and reportable range of the Tosoh T-uptake assay when performed on the A1A analyzer.