Orders LinkContactUsLinkComponent
Linearity Tumor Markers, AUDIT Microcontrols
Linearity Tumor Markers, AUDIT Microcontrols
Supplier:   Audit Microcontrols
Linearity Tumor Markers, AUDIT Microcontrols
Certificates
Linearity Tumor Markers, AUDIT Microcontrols
Linearity Tumor Markers, AUDIT Microcontrols
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Linearity FD HE4, HER2 tumor markers, Linearity FD tumor markers, and Linearity FD tumor markers for Siemens Centaur are assayed quality control materials consisting of five levels.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Product Family Options
Product Information
  • Type
    Analyte
    Stability
    Size
    Availability
    Price
  • 76359-704
    Linearity FD HE4, HER2 tumor markers
    Linearity FD HE4, HER2 tumor markers
    Freeze dried
    HE4, HER2
    7 days when stored at 2...8 °C
    5×1 ml
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 97068-934
    Linearity FD tumor markers
    Linearity FD tumor markers
    Freeze dried
    AFP, CA 125, CA 19-9, CA, 27-29, CEA, Total PSA, CA 15-3, Free PSA
    7 days when stored at 2...8 °C
    10×1 ml
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $915.37
    $915.37
    Pack of 5
       $915.37
    Supplier Number: K719M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 97068-936
    Linearity FD tumor markers for Siemens Centaur
    Linearity FD tumor markers for Siemens Centaur
    Freeze dried
    AFP, CA 125, CA 19-9, CA, 27-29, CEA, PSA, CA 15-3
    7 days when stored at 2...8 °C
    10×1 ml
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $922.43
    $922.43
    Pack of 5
       $922.43
    Supplier Number: K805M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.