Orders LinkContactUsLinkComponent
Linearity Thyroid, AUDIT Microcontrols
Linearity Thyroid, AUDIT Microcontrols
Supplier:   Audit Microcontrols
Linearity Thyroid, AUDIT Microcontrols
Certificates
Linearity Thyroid, AUDIT Microcontrols
Linearity Thyroid, AUDIT Microcontrols
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Linearity FD Thyroid for Abbott Architect I Series, Linearity FLQ Thyroid for Roche systems and Linearity FD Thyroid for Siemens Centaur are assayed quality control materials consisting of five levels.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Product Family Options
Product Information
  • Type
    Analyte
    Stability
    Size
    Availability
    Price
  • 76359-714
    Linearity FD Thyroid for Abbott Systems
    Linearity FD Thyroid for Abbott Systems
    Freeze dried
    Total T3, Total T4, Free T3, Free T4
    5 days when stored at 2...8 °C
    10×2 ml
    Log in for availability
    Log in for availability
    Kit of 5 (2ml)
    Kit of 5 (2ml) - $78.59
    $78.59
    Kit of 5 (2ml)
       $78.59
    Supplier Number: K832M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 76359-740
    Linearity FLQ Thyroid for Roche Systems
    Linearity FLQ Thyroid for Roche Systems
    Frozen liquid
    Total T3, Total T4, TSH, Free T3, Free T4
    7 days when stored at 2...8 °C
    10×2 ml
    Log in for availability
    Log in for availability
    Kit of 5 (2ml)
    Kit of 5 (2ml) - $352.67
    $352.67
    Kit of 5 (2ml)
       $352.67
    Supplier Number: K926M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 97068-944
    Linearity FD Thyroid for Siemens Centaur
    Linearity FD Thyroid for Siemens Centaur
    Freeze dried
    Total T3, Total T4, TSH, Free T3, Free T4
    7 days when stored at 2...8 °C
    10×3 ml
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $155.67
    $155.67
    Pack of 10
       $155.67
    Supplier Number: K807M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.