Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Linearity RF/CRP, AUDIT Microcontrols

Linearity RF/CRP, AUDIT Microcontrols

Linearity LQ RF/CRP, Liquid, CRP, RF, 5 days when stored at 2...8 °C, 5×1 ml
Catalog # 97068-906
Supplier:   Audit Microcontrols
Supplier #:  K720M-5
Linearity RF/CRP, AUDIT Microcontrols
Catalog # 97068-906
CAS Number:  
Linearity RF/CRP, AUDIT Microcontrols
Linearity RF/CRP, AUDIT Microcontrols

Linearity LQ RF/CRP, Liquid, CRP, RF, 5 days when stored at 2...8 °C, 5×1 ml

CAS Number:  
Price
$616.64
Log in for availability
Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

RF/CRP Linearity is assayed quality control material consisting of five levels of human based serum.

Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.

Specifications

Specifications

  • Description:
    Linearity LQ RF/CRP
  • Size:
    5×1 ml
  • Stability:
    5 days when stored at 2...8 °C
  • Analyte:
    CRP, RF
  • Type:
    Liquid

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.