Linearity Lipids, AUDIT Microcontrols
About this item
Universal linearity lipids, linearity lipids for Roche Systems, and linearity lipids for Beckman AU are assayed quality control materials consisting of five levels of bovine based serum albumin.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Universal linearity lipids, linearity lipids for Roche Systems, and linearity lipids for Beckman AU are assayed quality control materials consisting of five levels of bovine based serum albumin.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Product Family Options
Product Information
- TypeAnalyteStabilitySizeAvailabilityPrice
- 10672-490Linearity FD lipids for Roche SystemsLinearity FD lipids for Roche SystemsFreeze driedApolipoprotein A-1, Apolipoprotein B, Cholesterol-HDL, Cholesterol-LDL, Cholesterol, Triglycerides5 days when stored at 2...8 °C5×2 mlLog in for availabilityEach(2ml)$738.40Supplier Number: K882M-5Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 76359-712Linearity FD lipids for Beckman AULinearity FD lipids for Beckman AUFreeze driedCholesterol-HDL, Cholesterol-LDL, Cholestero, Triglycerides5 days when stored at 2...8 °C5×2 mlLog in for availabilityKit of 5 (2ml)$284.21Supplier Number: K826M-5Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 97068-896Linearity FD lipidsLinearity FD lipidsFreeze driedApolipoprotein A-1, Apolipoprotein B, Cholesterol-HDL, Cholesterol-LDL, Cholesterol, Triglycerides5 days when stored at 2...8 °C5×2 mlLog in for availabilityPack of 5$614.29Supplier Number: K709M-5Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.