Orders LinkContactUsLinkComponent
Linearity Immunoassay, AUDIT Microcontrols
Linearity Immunoassay, AUDIT Microcontrols
Supplier:   Audit Microcontrols
Linearity Immunoassay, AUDIT Microcontrols
Certificates
Linearity Immunoassay, AUDIT Microcontrols
Linearity Immunoassay, AUDIT Microcontrols
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Linearity FD immunoassay and Linearity FD Immunoassay for Abbott Architect I series are assayed quality control materials consisting of five levels.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Product Family Options
Product Information
  • Type
    Analyte
    Stability
    Size
    Availability
    Price
  • 76359-716
    Linearity FD immunoassay for Abbott Architect I Series
    Linearity FD immunoassay for Abbott Architect I Series
    Freeze dried
    CEA, Ferritin, Folate, FSH, LH, Progesterone, Prolactin, Free PSA, Total PSA, Testosterone, TSH, Vitamin B12
    5 days when stored at 2...8 °C
    5×5 ml
    Log in for availability
    Log in for availability
    Kit of 5 (5ml)
    Kit of 5 (5ml) - $2,211.56
    $2,211.56
    Kit of 5 (5ml)
       $2,211.56
    Supplier Number: K833M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 97068-904
    Linearity FD immunoassay
    Linearity FD immunoassay
    Freeze dried
    Cortisol, Digoxin, Estradiol, Ferritin, Folate, FSH, hCG, LH, Progesterone, Prolactin, PSA, T3-Free, T3-Total, T4-Free, T4-Total, Testosterone, TSH, Vitamin B12
    5 days when stored at 2...8 °C
    10×5 ml
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $642.30
    $642.30
    Pack of 5
       $642.30
    Supplier Number: K714M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.