Orders LinkContactUsLinkComponent
Linearity D-Dimer, AUDIT Microcontrols
Linearity D-Dimer, AUDIT Microcontrols
Supplier:   Audit Microcontrols
Linearity D-Dimer, AUDIT Microcontrols
Certificates
Linearity D-Dimer, AUDIT Microcontrols
Linearity D-Dimer, AUDIT Microcontrols
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Universal linearity D-Dimer, linearity D-Dimer for bioMerieux VIDAS and miniVIDAS, linearity D-Dimer for PATHFAST Immunoanalyzer, linearity D-Dimer for Siemens Stratus are assayed quality control materials consisting of five levels of human plasma.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Product Family Options
Product Information
  • Type
    Analyte
    Stability
    Size
    Availability
    Price
  • 76359-720
    Linearity FD D-Dimer for bioMerieux VIDAS and miniVIDAS
    Linearity FD D-Dimer for bioMerieux VIDAS and miniVIDAS
    Freeze dried
    D-Dimer
    2 days when stored at 2...8 °C
    5×1 ml
    Log in for availability
    Log in for availability
    Kit of 5 (1ml)
    Kit of 5 (1ml) - $3,176.22
    $3,176.22
    Kit of 5 (1ml)
       $3,176.22
    Supplier Number: K845M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 76359-722
    Linearity FD D-Dimer for PATHFAST immunoanalyzer
    Linearity FD D-Dimer for PATHFAST immunoanalyzer
    Freeze dried
    D-Dimer
    24 hours when stored at 2...8 °C
    5×1 ml
    Log in for availability
    Log in for availability
    Kit of 5 (1ml)
    Kit of 5 (1ml) - $568.68
    $568.68
    Kit of 5 (1ml)
       $568.68
    Supplier Number: K883M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 76359-730
    Linearity FD D-Dimer for Siemens Stratus
    Linearity FD D-Dimer for Siemens Stratus
    Freeze dried
    D-Dimer
    2 days when stored at 2...8 °C
    5×1 ml
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 97068-942
    Linearity FD D-Dimer
    Linearity FD D-Dimer
    Freeze dried
    D-Dimer
    2 days when stored at 2...8 °C
    5×1 ml
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $494.86
    $494.86
    Pack of 5
       $494.86
    Supplier Number: K717M-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.