Order Entry
Puerto Rico
ContactUsLinkComponent
Molecular sieve 3A (0.3 nm, 3 Å) absorption tube for H₂O, with indicator, Supelco®
Molecular sieve 3A (0.3 nm, 3 Å) absorption tube for H₂O, with indicator, Supelco®
Catalog # EM1.06107.0003
Supplier:  MilliporeSigma
CAS Number:  
Molecular sieve 3A (0.3 nm, 3 Å) absorption tube for H₂O, with indicator, Supelco®
Catalog # EM1.06107.0003
Supplier:  MilliporeSigma
Supplier Number:  1.06107.0003
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic tube
  • Size:
    3 tubes
  • Cat. No.:
    EM1.06107.0003
  • Supplier no.:
    1.06107.0003

Specifications

Specification Test Results

  • Appearance:
    passes test

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:308080-99-1
 :1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613