Order Entry
Puerto Rico
ContactUsLinkComponent
 
Digi-Sense® Professional Dual-Laser Infrared Thermometer with Bluetooth® Connectivity, Cole-Parmer
Catalog # 76360-048
Supplier:  Antyila Scientific
undefined
Digi-Sense® Professional Dual-Laser Infrared Thermometer with Bluetooth® Connectivity, Cole-Parmer
Catalog # 76360-048
Supplier:  Antyila Scientific
Supplier Number:  20250-25

Specifications

  • Description:
    Digi-Sense® professional dual-laser infrared thermometer with Bluetooth® connectivity
  • Distance to spot ratio:
    50:1
  • Cat. No.:
    76360-048

Specifications

About this item

View, store, and share real-time data on your smartphone.

  • Get the most out of your data with our user-friendly mobile app
  • Unlimited data storage based on smart device capacity
  • Reliable measurements, guaranteed, precalibrated to NIST-traceable standards
  • Internal data logging saves up to 32K IR readings
  • External type K thermocouple input for contact measurements
  • Dual-laser sighting to quickly pinpoint target areas
  • Backlight dot-matrix display permits viewing in low-light conditions

Using our free Digi-Sense D/S IR Therm app, this dual-laser infrared (IR) thermometer with Bluetooth® technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your work—ideal for record keeping at large sites.

The IR thermometer offers fast response time and high accuracy due to its 50 to 1 distance-to-sight ratio. Dual targeting lasers provide precise indication of the measurement area. It is ideal for a wide range of applications including food preparation, safety and fire inspection, plastic molding, marine, HVAC, and fleet maintenance.

Advanced features include internal data logging, high/low temperature alarms, adjustable emissivity, temperature differential, data Hold, Max/Min/Avg readings, USB interface for data transmission, and auto power-off. The meter also supports contact measurement through a type K thermocouple input.

The instrument is fully tested and calibrated to NIST-traceable standards for reliable measurements right out of the box, saving you time and money!

Certifications: NIST-traceable calibration report.
Delivery: Supplied with type K flexible probe, software, tripod, carrying case, 9 V battery, and NIST-traceable calibration report with data.