Order Entry
Puerto Rico
ContactUsLinkComponent
Linearity Blood Gas, AUDIT Microcontrols
Linearity Blood Gas, AUDIT Microcontrols
Catalog # 97068-912
CAS Number:  
undefined
Linearity Blood Gas, AUDIT Microcontrols
Catalog # 97068-912
Supplier Number:  K727M-5
CAS Number:  

Specifications

  • Description:
    Cal Ver LQ blood gas
  • Size:
    15×2 ml
  • Cat. No.:
    97068-912
  • Stability:
    60 Seconds
  • Analyte:
    BUN, Calcium-Ionized, Chloride, Creatinine, Glucose, Lactate (Lactic Acid), pCO2, pH, pO2, Potassium, Sodium
  • Type:
    Liquid
  • Supplier no.:
    K727M-5

Specifications

About this item

Blood gas calibration verification is assayed quality control material consisting of five levels of buffers and salts in a bovine based material and equilibrated with known levels of carbon dioxide (CO₂), oxygen (O₂), and nitrogen.

A quality control material intended for use in the quantitative verification of calibration and reportable range.