KIMBLE® Soxhlet Extraction Body, DWK Life Sciences
Catalog # KT586280-0022
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Soxhlet extraction body
- Height:300 mm
- Cat. No.:KT586280-0022
- Fits:Thimble size 0022/0222
- Supplier no.:586280-0022
Specifications
About this item
Extractor with PTFE stopcock outlet for periodic solvent removal.
- Stopcock has an PTFE plug
- Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.