KIMBLE® Soxhlet/Dean Stark Extraction Apparatus, DWK Life Sciences
Catalog # KT585150-0023
Supplier: Avantor
Specifications
- Description:Soxhlet/dean stark extraction apparatus
- Environmentally Preferable:
- Cat. No.:KT585150-0023
- Supplier no.:585150-0023
Specifications
About this item
This apparatus combines two analytical techniques, Soxhlet extraction and Dean-Stark azeotropic distillation, into a single process for the removal of water, chlorinated dibenzo-p-dioxins (CCDs) and chlorinated dibenzofurans (CDFs) from wet particulate samples.
- Determination of water and CDDs/ CDFs on the same sample aliquot is achieved without the need for desiccation prior to soxhlet extraction
- Loss of analytes and sample contamination are avoided
- Manufactured from 33 expansion, low extractable borosilicate glass
Uses a KT586500-0233 fritted glass extraction thimble, not supplied. PTFE stopcock plug is KT821001-0002.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.