ContactUsLinkComponent
KIMBLE® Micro Distilling Column, DWK Life Sciences
Catalog # KT569261-0319
undefined
KIMBLE® Micro Distilling Column, DWK Life Sciences
Catalog # KT569261-0319
Supplier Number:  569261-0319
Specifications
  • Description:
    Micro distilling column
  • Length:
    122 mm
  • Cat. No.:
    KT569261-0319
  • [ST] joint size:
    19/22
  • Supplier no.:
    569261-0319
Specifications
About this item
Micro-type distilling column designed to prevent flooding while retaining residue material.
  • Manufactured from 33 expansion, low extractable borosilicate glass
Kimble® KT569261-0319 micro distilling column with three chambers and 19/22 [ST] joints, no hooks, length: 122 mm
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.