KIMBLE® Drying Tubes, DWK Life Sciences
KT562500-2440
- [ST] joint no.:24/40
- Description:Drying tube
- Cat. No.:KT562500-2440
- Overall length:185 mm
- Supplier no.:562500-2440
U-shaped drying tube.
- Inner joint at the bottom with a bulb desiccant chamber
- Manufactured from 33 expansion, low extractable borosilicate glass
Joint length: full, overall height: 185 mm, overall width: 80 mm
: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
: Supplied with a bent inlet tube.