Order Entry
Puerto Rico
ContactUsLinkComponent
 
KIMBLE® KONTES® Dewar Condenser with Hose Connection Sidearm, DWK Life Sciences
  KT457500-2420
undefined
KIMBLE® KONTES® Dewar Condenser with Hose Connection Sidearm, DWK Life Sciences
  KT457500-2420
 :  457500-2420

 

  • [ST] joint:
    24/40
  • Description:
    Dewar condenser, 200 mm depth
  • Height:
    295 mm
  • Cat. No.:
    KT457500-2420
  • Fits:
    ⅜" ID tubes
  • Supplier no.:
    457500-2420

 

 

Sidearm is a hose connection for attachment to flexible tubing

  • Large top opening for easy introduction of coolant mixtures such as solid carbon dioxide/alcohol, dry ice/alcohol, dry ice, or liquid nitrogen
  • Manufactured from 33 expansion, low extractable borosilicate glass

Lower joint will work with polyacetal standard taper clamps.

 : Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.