KIMBLE® Coil-Type Reflux Condenser with Two Upper Hose Barbs and Angled Outer Joint, DWK Life Sciences
KT457050-0000
- [ST] joint:24/40
- Description:Reflux condenser
- Height:370 mm
- Cat. No.:KT457050-0000
- Condensing area:261 cm²
- Supplier no.:457050-0000
Coil-type condenser for greater cooling capacity.
- Hook above lower standard taper joint for use with a KT675500-3445 KEM-KLAMP™
- Manufactured from 33 expansion, low extractable borosilicate glass
KT675000-0028, KT675000-0040 polyacetal standard taper clamps may also be used.
: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.