KIMBLE® TLC Reagent Sprayers with Standard Taper Joint, DWK Life Sciences
Catalog # KT422550-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:5 ml
- Description:Reagent sprayer
- Environmentally Preferable:
- Cat. No.:KT422550-0000
- [ST] joint no.:14/20
- Supplier no.:422550-0000
Specifications
About this item
These reagent sprayers have an all-glass construction which makes them completely steam autoclavable.
- Operate between 3 and 5 psi (0.21 to 0.35 kg/cm²)
- Manufactured from 33 expansion, low extractable borosilicate glass
This TLC reagent sprayer with standard ground joints provides uniform spray mist of visualization reagents for TLC and paper chromatography. Use with all standard TLC reagents and most corrosive liquids. Includes test tube and flask.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.