Anti-IgG Sheep Polyclonal Antibody 500 µg
76334-050
- Antibody Type:Primary
- Antigen Name:No antigen
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Sheep
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:500 µL
- Form:Lyophilized
- Shipping Temperature:Room (ambient) temperature
- Purification:Whole serum
- Cat. No.:76334-050
Sheep serum from non-immunized animals was purified by protein G affinity chromatography. This sheep IgG fraction can be used as negative control antibody for various immunological techniques where sheep primary antibodies are used.
Applications: Use as negative control for IgG-purified sheep primary antibodies and at matching concentration.
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: