Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-BDNF, NGF, NTF3, NTF4 Sheep Polyclonal Antibody
  76333-986
 :  Biosensis
Anti-BDNF, NGF, NTF3, NTF4 Sheep Polyclonal Antibody
  76333-986
 :  Biosensis
 :  KIT-NT-500

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Brain-derived neurotrophic factor, Beta-nerve growth factor, Neurotrophin-3, Neurotrophin-4
  • Antigen Symbol:
    BDNF, NGF, NTF3, NTF4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    5 × 500 µg
  • Neutralisation:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    BDNF, Ngf, NTF3, NTF4
  • Storage Temperature:
    Store lyophilised, unopened vials at 2…8° C or lower. After reconstitution, prepare aliquots and store at -20° C for a higher stability. Avoid freeze-thaw cycles.
  • Shipping Temperature:
    Room (ambient) temperature
  • Immunogen:
    BDNF: recombinant human BDNF
    NGF: native mouse beta NGF purified from submaxillary salivary gland (95% purity by PAGE)
    NT3: recombinant human NT3
    NT4/5: recombinant human NT4
  • Purification:
    Protein G purified.
  • Cat. No.:
    76333-986

 

 

Neurotrophins are growth factors that regulate neuronal development, maintenance and plasticity. These homo-dimeric proteins activate a multitude of intracellular signalling cascades through their interaction with Trk receptors and p75NTR.

The Neurotrophin Antibody Kit contains a collection of Biosensis' popular sheep antibodies raised against BDNF (S-015-500), NGF (S-052-500), NT3 (S-057-500) and NT4/5 (S-059-500). The kit is completed with one vial of normal sheep IgG (S-1754-500) for use as negative control. This full size version of Biosensis' Neurotrophin Antibody Kit contains 500 ug sheep IgG of each antibody.

This kit presents a cost-effective way to investigate neurotrophin protein expression and function by immunological techniques such as immunohistochemistry, ELISA, and inhibition of biological activity.

Applications: Immunohistochemistry: 1-10 ug/mL. BDNF antibody works best on Zamboni's fixed, frozen tissue, and is not recommended for paraffin-embedded tissue. ELISA: 1-10 ug/mL. Western Blotting: 1-10 ug/mL. Western blotting is not recommended for the BDNF antibody.
Inhibition of biological activity in vitro/in vivo: Expected ED50 (in vivo) values are in the range of 2-10 ug/mL. The NGF antibody completely inhibits neuronal survival and the outgrowth actions of murine NGF in chicken DRG in vitro. Biosensis recommends optimal dilutions/concentrations should be determined by the end user. The negative control (normal sheep IgG) should be used at the same concentration as the neurotrophin antibodies.

Type: Primary
Antigen: BDNF, NGF, NTF3, NTF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, rat, mouse