About this item
CELLvo™ human mesenchymal stem cells-Wharton's Jelly are adult multipotent stem cells cryopreserved at passage two.
- Highly proliferative
- Smaller cell size
- Xeno-free
- Higher percentage of SSEA4+ cells
- >20 population doublings
Wharton's Jelly is a rich source of highly proliferative, multipotent, mesenchymal stem cells. These cells are known to be highly immunomodulatory and be capable of trilineage differentiation.
CELLvo™ human mesenchymal stem cells- Wharton's Jelly have been isolated and cultured under xeno free conditions on CELLvo™ XF-Matrix. CELLvo™ MSC-WJ proliferate more quickly, have a smaller cell size and show a higher percentage of SSEA4 (+) cells as compared to Wharton's Jellly MSCs isolated and cultured on other substrates.
Thaw cells quickly in a 37 °C waterbath. Approximate thaw time for a 1 ml sample is 2 to 3 minutes. Do not allow the cell vial to warm above chilled temperatures. Dilute cell/CryoStor® mixture in an appropriate volume of cell culture media and plate cells in the appropriate configuration. Live/dead fluorescsent assays or metabolic assays 24 hours post-thaw are recommended for accurate assessment of cell survival.
CELLvo™ products are not for human use. These products should not be used for diagnostic or therapeutic purposes. Handle in accordance with established bio-safety practices.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Conditions:Upon arrival, store cells in liquid nitrogen vapor
- Storage Buffer:CryoStor®CS5
- Tested Applications:Multilineage differentiation, flow cytometry and viability