



Dichloroacetic acid (DCA) 10% in anhydrous toluene deblocking reagent, for biosynthesis, VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
CAS Number:79-43-6
:
Formula:C₂H₂Cl₂O₂
MDL Number:MFCD00004223
ADR:3 (8),II
:128.94 g/mol
UN:2924
:GHS02
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Description (Clear liquid free of suspended matter):Passes test
- Dichloroacetic acid:9.5 to 10.5 % Vol
- Color (at time of packaging):Max 10 APHA
- Water (by KF coulometry) :Max 150 ppm
- Dichloroacetic acid (component purity) :Min 98.0 %
- Toluene (component purity) :Min 99.9 %
- Titrable Acid or Base:89.5 to 90.5 %