Sulphuric acid 0.5 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:3264
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS05
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Amount-of-substance concentration:0.4975 - 0.5025 mol/L
- Form:liquid
- Measurement uncertainty:± 0.0015 mol/L
Specification Test Results
Related Information
Certificates
Search by Lot NumberAYN4_Analytical_Chemistry_-Electrochemistry EMD Millipore Sustainable Products Brochure MilliporeSigma Titripac Packaging MilliporeSigma Titripac® The Clever Packaging Solution
Product Literature
spec_Sulfuric acid solution-0.5M Stability data sheet Titripac_109072
Product specification
VWR GHS Compliance Letter (US)