Sodium hydroxide 0.02 mol/l (0.02 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # EM1.09142.0500
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:500 ml
- Cat. No.:EM1.09142.0500
- Supplier no.:1.09142.0500
Specifications
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.01990 - 0.02002
- Measurement uncertainty:+ / - 0.00006
- Traceability:NIST SRM
Specification Test Results
About this item
:
:40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:215-185-5
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548