



Potassium hydroxide 2 mol/l (2 N) in methanol, Titripur® volumetric solution, Supelco®
Catalog # EM1.11787.2500
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:EM1.11787.2500
- Supplier no.:1.11787.2500
Specifications
About this item
:
:56.11 g/mol
ADR:3 (6.1, 8),II
:M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:3286
:GHS02
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553