Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®
Catalog # EM1.09351.1000
Supplier:  MilliporeSigma
CAS Number:  
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®
Catalog # EM1.09351.1000
Supplier:  MilliporeSigma
Supplier Number:  1.09351.1000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    EM1.09351.1000
  • Supplier no.:
    1.09351.1000

Specifications

About this item

 :
 :56.11 g/mol
ADR:3 (6.1, 8),II
 :M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:3286
 :GHS02
 :GHS06
 :GHS05
 :GHS08
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553